ENA311760369 Enamine

N-(pyrimidin-2-yl)piperidine-4-carboxamide dihydrochloride

  • MDL number MFCD18089497
  • Cas Number : 1308646-67-4
  • Molecular Weight : 279.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72397
Availability details for
Quantity
  • Availability
  • Add to Cart