ENA311760370 Enamine

2-(diethylamino)-3-methylbutanoic acid hydrobromide

  • MDL number MFCD18089499
  • Cas Number : 1485774-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Product Link https://www.enaminestore.com/catalog/EN300-72405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72405
Availability details for
Quantity
  • Availability
  • Add to Cart