ENA311760378 Enamine

2-{[1-(2-chloro-4-fluorophenyl)ethyl]amino}-N-methylacetamide hydrochloride

  • MDL number MFCD18089508
  • Cas Number : 1305712-17-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72472
Availability details for
Quantity
  • Availability
  • Add to Cart