ENA311760379 Enamine

4,4,7-trimethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD18089509
  • Cas Number : 1305711-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72473
Availability details for
Quantity
  • Availability
  • Add to Cart