ENA311760380 Enamine

1-cyclohexyl-4-(piperidin-3-yl)piperazine

  • MDL number MFCD18089510
  • Cas Number : 1308650-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72477
Availability details for
Quantity
  • Availability
  • Add to Cart