ENA311760381 Enamine

[(4-fluorophenyl)(pyridin-3-yl)methyl](propyl)amine dihydrochloride

  • MDL number MFCD18089512
  • Cas Number : 1305711-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-72495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72495
Availability details for
Quantity
  • Availability
  • Add to Cart