ENA311760383 Enamine

1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD18089514
  • Cas Number : 1305711-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72498
Availability details for
Quantity
  • Availability
  • Add to Cart