ENA311760388 Enamine

1-(3-chloropyridin-2-yl)-5-(difluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD18089519
  • Cas Number : 1305711-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72515
Availability details for
Quantity
  • Availability
  • Add to Cart