ENA311760389 Enamine

2-{5,7-dimethyl-2-oxo-1H,2H,4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidin-6-yl}acetic acid

  • MDL number MFCD18089520
  • Cas Number : 1305710-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72522

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72522
Availability details for
Quantity
  • Availability
  • Add to Cart