ENA311760391 Enamine

2-methyl-1-(1-methyl-1H-imidazol-2-yl)propan-1-one

  • MDL number MFCD18089522
  • Cas Number : 138536-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72536
Availability details for
Quantity
  • Availability
  • Add to Cart