ENA311760394 Enamine

2-[4-(pyridin-2-yl)-1,3-thiazol-2-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD18089525
  • Cas Number : 1308650-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72557
Availability details for
Quantity
  • Availability
  • Add to Cart