ENA311760617 Enamine

3-[4-(propan-2-yl)benzoyl]pyridine

  • MDL number MFCD12094682
  • Cas Number : 61780-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72116
Availability details for
Quantity
  • Availability
  • Add to Cart