ENA311760625 Enamine

1-(4-bromophenyl)-3-phenylpropane-1,3-dione

  • MDL number MFCD11539406
  • Cas Number : 25856-01-3
  • Molecular Weight : 303.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-93
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72136

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-72136
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart