ENA311760626 Enamine

2-(3-hydroxypropyl)-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one

  • MDL number MFCD12186713
  • Cas Number : 53689-26-2
  • Molecular Weight : 193.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72137
Availability details for
Quantity
  • Availability
  • Add to Cart