ENA311760633 Enamine

3-[1-(3-chloro-4-methylphenyl)-2,5-dioxoimidazolidin-4-yl]propanoic acid

  • MDL number MFCD13744132
  • Cas Number : 1192227-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 193...195
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72148
Availability details for
Quantity
  • Availability
  • Add to Cart