ENA311760634 Enamine

2-(aminomethyl)-N-ethyl-N-methylaniline

  • MDL number MFCD11622588
  • Cas Number : 1094800-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71634
Availability details for
Quantity
  • Availability
  • Add to Cart