ENA311760646 Enamine

2-amino-N,N-diethylethane-1-sulfonamide

  • MDL number MFCD03077759
  • Cas Number : 92739-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71635
Availability details for
Quantity
  • Availability
  • Add to Cart