ENA311760650 Enamine

[4-(4-ethylpiperazin-1-yl)-3-fluorophenyl]methanamine

  • MDL number MFCD11191697
  • Cas Number : 1039892-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71636

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71636
Availability details for
Quantity
  • Availability
  • Add to Cart