ENA311760652 Enamine

tert-butyl 3-(2-aminoethyl)piperidine-1-carboxylate

  • MDL number MFCD04038458
  • Cas Number : 259180-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71641
Availability details for
Quantity
  • Availability
  • Add to Cart