ENA311760657 Enamine

2-amino-5-(pyrrolidin-1-yl)benzoic acid

  • MDL number MFCD12816217
  • Cas Number : 159526-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71649
Availability details for
Quantity
  • Availability
  • Add to Cart