ENA311760663 Enamine

2-(3-chloro-4-fluorophenoxy)-2-phenylacetic acid

  • MDL number MFCD12542597
  • Cas Number : 1097168-10-9
  • Molecular Weight : 280.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72193
Availability details for
Quantity
  • Availability
  • Add to Cart