ENA311760665 Enamine

2-amino-5-(4-methylpiperazin-1-yl)benzoic acid

  • MDL number MFCD12869794
  • Cas Number : 835879-06-6
  • Molecular Weight : 235.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71651
Availability details for
Quantity
  • Availability
  • Add to Cart