ENA311760670 Enamine

ethyl[(1-methyl-1H-imidazol-2-yl)(pyridin-3-yl)methyl]amine

  • MDL number MFCD14623557
  • Cas Number : 1250392-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72209

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72209
Availability details for
Quantity
  • Availability
  • Add to Cart