ENA311760678 Enamine

2-(2-phenylethyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD14539677
  • Cas Number : 65346-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72220

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72220
Availability details for
Quantity
  • Availability
  • Add to Cart