ENA311760679 Enamine

methyl 2-phenyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD15142831
  • Cas Number : 172678-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72221
Availability details for
Quantity
  • Availability
  • Add to Cart