ENA311760681 Enamine

2-amino-1-(thiophen-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD00159551
  • Cas Number : 70996-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215...217
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72223

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-72223
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart