ENA311760682 Enamine

5-(1-phenylpropyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09808094
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72224
Availability details for
Quantity
  • Availability
  • Add to Cart