ENA311760683 Enamine

5-(3-phenylpropyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD10693883
  • Cas Number : 1152585-28-8
  • Molecular Weight : 203.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72225
Availability details for
Quantity
  • Availability
  • Add to Cart