ENA311760684 Enamine

3-[1-(3-methylphenyl)-2,5-dioxoimidazolidin-4-yl]propanoic acid

  • MDL number MFCD12691255
  • Cas Number : 1173293-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72227
Availability details for
Quantity
  • Availability
  • Add to Cart