ENA311760686 Enamine

3-[1-(4-chlorophenyl)-2,5-dioxoimidazolidin-4-yl]propanoic acid

  • MDL number MFCD12691256
  • Cas Number : 1173293-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72229

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72229
Availability details for
Quantity
  • Availability
  • Add to Cart