ENA311760687 Enamine

5-bromo-3-fluoro-2-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD17012834
  • Cas Number : 612541-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72230
Availability details for
Quantity
  • Availability
  • Add to Cart