ENA311760689 Enamine

2-(2-hydroxyethyl)-2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one

  • MDL number MFCD12181095
  • Cas Number : 56654-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72232

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72232
Availability details for
Quantity
  • Availability
  • Add to Cart