ENA311760698 Enamine

1-(1-chloroethyl)-4-(trifluoromethyl)benzene

  • MDL number MFCD11186465
  • Cas Number : 85289-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72247
Availability details for
Quantity
  • Availability
  • Add to Cart