ENA311760704 Enamine

5-bromo-2-(1H-pyrazol-1-yl)pyridin-3-amine

  • MDL number MFCD14600568
  • Cas Number : 1249484-03-4
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72253
Availability details for
Quantity
  • Availability
  • Add to Cart