ENA311760708 Enamine

2-amino-1-(2-chlorophenyl)ethan-1-one hydrochloride

  • MDL number MFCD09787499
  • Cas Number : 16442-79-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-172
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72261

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72261
Availability details for
Quantity
  • Availability
  • Add to Cart