ENA311760712 Enamine

methyl 5-(chlorosulfonyl)furan-2-carboxylate

  • MDL number MFCD02180740
  • Cas Number : 69816-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72267

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-72267
s_phrases_eu 53-23-24/25-36/37
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart