ENA311760715 Enamine

3-methanesulfonylthiophene-2-carboxylic acid

  • MDL number MFCD18089406
  • Cas Number : 60166-82-7
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-71660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71660
Availability details for
Quantity
  • Availability
  • Add to Cart