ENA311760717 Enamine

4-(pyrrolidin-1-yl)butan-2-amine

  • MDL number MFCD09740583
  • Cas Number : 138548-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71662
Availability details for
Quantity
  • Availability
  • Add to Cart