ENA311760719 Enamine

4-(benzyloxy)-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD18089408
  • Cas Number : 211805-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71663

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71663
Availability details for
Quantity
  • Availability
  • Add to Cart