ENA311760722 Enamine

2-(3,5-dimethyl-1H-pyrazol-4-yl)propanoic acid

  • MDL number MFCD08700814
  • Cas Number : 1093416-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72285
Availability details for
Quantity
  • Availability
  • Add to Cart