ENA311760729 Enamine

ethyl[(4-methoxyphenyl)(pyridin-3-yl)methyl]amine

  • MDL number MFCD12419670
  • Cas Number : 1153032-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72295
Availability details for
Quantity
  • Availability
  • Add to Cart