ENA311760736 Enamine

4-(2-methylpropoxy)-1-phenyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD12446029
  • Cas Number : 1152512-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72309
Availability details for
Quantity
  • Availability
  • Add to Cart