ENA311760745 Enamine

ethyl({[4-(propan-2-yl)phenyl](pyridin-3-yl)methyl})amine

  • MDL number MFCD12589261
  • Cas Number : 1154165-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-72325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72325
Availability details for
Quantity
  • Availability
  • Add to Cart