ENA311760753 Enamine

4-isobutyl-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD16068932
  • Cas Number : 1311416-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72340
Availability details for
Quantity
  • Availability
  • Add to Cart