ENA311760754 Enamine

1-(4-acetylpiperazin-1-yl)-3-aminopropan-1-one hydrochloride

  • MDL number MFCD06796421
  • Cas Number : 917758-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72364

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72364
Availability details for
Quantity
  • Availability
  • Add to Cart