ENA311760758 Enamine

2-(4-bromo-2-fluorophenoxy)-2-phenylacetic acid

  • MDL number MFCD12556929
  • Cas Number : 1040314-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71697

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71697
Availability details for
Quantity
  • Availability
  • Add to Cart