ENA311760762 Enamine

1-(2,4-dichlorophenyl)-4-methylpentan-2-one

  • MDL number MFCD11920578
  • Cas Number : 1177228-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72375
Availability details for
Quantity
  • Availability
  • Add to Cart