ENA311760766 Enamine

[1-(4-methoxypiperidin-1-yl)cyclopentyl]methanamine

  • MDL number MFCD12167165
  • Cas Number : 1096355-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-72381
Availability details for
Quantity
  • Availability
  • Add to Cart