ENA311760773 Enamine

1-[4-methyl-2-(4-methylphenyl)-1,3-thiazol-5-yl]ethan-1-amine

  • MDL number MFCD09882220
  • Cas Number : 1017379-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-72392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72392
Availability details for
Quantity
  • Availability
  • Add to Cart