ENA311760777 Enamine

1-(1-phenylcyclobutanecarbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD13668065
  • Cas Number : 1099048-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-72401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72401
Availability details for
Quantity
  • Availability
  • Add to Cart